missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-4-piperidinyl)methanamine, 97%

Product Code. 15249746 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15249746 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15249746 Supplier Thermo Scientific Acros Supplier No. 451460100
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7149-42-0 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7149-42-0
Boiling Point 80.0°C to 81.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C7H16N2
MDL Number MFCD05022430
Quantity 10 g
Synonym 1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine
InChI Key AGTPSAZJSOQXHJ-UHFFFAOYSA-N
SMILES CN1CCC(CN)CC1
IUPAC Name (1-methylpiperidin-4-yl)methanamine
Molecular Weight (g/mol) 128.22
PubChem CID 81574
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-4-piperidinyl)methanamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (1-Methyl-4-piperidinyl)methanamine, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.