missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-phenylpiperidine Hydrochloride (MPP Hydrochloride), Mikromol™

Product Code. 30052275
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052275

Brand: Mikromol MM0315.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Methyl-4-phenylpiperidine Hydrochloride (MPP Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methyl-4-phenylpiperidine Hydrochloride
CAS 10272-50-1
Analyte or Component Names 1-Methyl-4-phenylpiperidine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H17 N . Cl H
Certifications/Compliance ISO 9001
Synonym Pethidine Hydrochloride Imp. A (EP), Pethidine Imp. A (EP), Piperidine, 1-methyl-4-phenyl-, hydrochloride (1:1), Piperidine, 1-methyl-4-phenyl-, hydrochloride (7CI,8CI,9CI), 1-Methyl-4-phenylpiperidine hydrochloride, N-Methyl-4-phenylpiperidine hydrochloride, Pethidine Imp. A (EP) as Hydrochloride, MPP HCl, Pethidine Hydrochloride Impurity A as Hydrochloride, Pethidine Impurity A as Hydrochloride
InChI Formula InChI=1S/C12H17N.ClH/c1-13-9-7-12(8-10-13)11-5-3-2-4-6-11;/h2-6,12H,7-10H2,1H3;1H
SMILES Cl.CN1CCC(CC1)c2ccccc2
IUPAC Name 1-methyl-4-phenylpiperidine;hydrochloride
Molecular Weight (g/mol) 211.73
Quantity 100 mg
Formula Weight 211.1128 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.