missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(2-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15400039
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
62208-63-3
C11H15N3O2
221.26
MFCD00156356
SSRLRVMFEGGGMQ-UHFFFAOYSA-N
1-methyl-4-2-nitrophenyl piperazine, piperazine, 1-methyl-4-2-nitrophenyl, cbmicro_037654, 1-methyl-4-2-nitro-phenyl-piperazine, 2-4-methylpiperazin-1-yl nitrobenzene, 2-4-methylpiperazin-1-yl-1-nitrobenzene
410640
1-methyl-4-(2-nitrophenyl)piperazine
CN1CCN(CC1)C1=CC=CC=C1[N+]([O-])=O
This item is not returnable. View return policy
62208-63-3
C11H15N3O2
221.26
MFCD00156356
SSRLRVMFEGGGMQ-UHFFFAOYSA-N
1-methyl-4-2-nitrophenyl piperazine, piperazine, 1-methyl-4-2-nitrophenyl, cbmicro_037654, 1-methyl-4-2-nitro-phenyl-piperazine, 2-4-methylpiperazin-1-yl nitrobenzene, 2-4-methylpiperazin-1-yl-1-nitrobenzene
410640
1-methyl-4-(2-nitrophenyl)piperazine
CN1CCN(CC1)C1=CC=CC=C1[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 221.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Methyl-4-(2-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'