missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™

Product Code. 10397202
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397202 1 g Amber Glass Bottle 1g
10246532 250 mg Amber Glass Bottle 250mg
11366815 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10397202 Supplier Thermo Scientific Maybridge Supplier No. AW00087DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 519056-67-8
Molecular Formula C5H4ClF3N2O2S
Molecular Weight (g/mol) 248.604
InChI Key HJFWSNIHZZKYIK-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794561
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.