missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride, 97+%, Thermo Scientific™

Product Code. p-4509979
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10391423 250 mg Amber Glass Bottle 250mg
10123652 1 g Amber Glass Bottle 1g
10709615 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10391423 Supplier Thermo Scientific Maybridge Supplier No. GK00779CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 126674-98-4
Molecular Formula C6H4ClF3N2O
Molecular Weight (g/mol) 212.556
InChI Key KFKVECZQALNWSR-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 1-methyl-3-trifluoromethyl, acmc-20ms45, 3-trifluoromethyl-1-methyl-1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethylpyrazole-4-carboxylic acid chloride, 1-methyl-4-trifluoromethyl-1h-pyrazole-3-carbonyl chloride, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794581
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-carbonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.56
Percent Purity 95%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.