missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Methyl-3-pyrrolidinone, 96%

Product Code. 15474038
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15474038

Brand: Alfa Aesar™ H57585.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68165-06-0
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD09991706
InChI Key SLPUTJFVMJPMKV-UHFFFAOYSA-N
Synonym 1-methyl-3-pyrrolidinone, 1-methyl-3-pyrrolidone, 3-pyrrolidone, 1-methyl, 3-pyrrolidinone, 1-methyl, pubchem20342, 1-methyl-pyrrolidin-3-one, ksc497q2t, 1methyl-3-pyrrolidinone, 1-methyl-1-azacyclopentan-4-one
PubChem CID 144283
IUPAC Name 1-methylpyrrolidin-3-one
SMILES CN1CCC(=O)C1

Specifications

Flash Point 42°C (108°F)
Quantity 1 g
UN Number UN1993
Formula Weight 99.13
Percent Purity 96%
Chemical Name or Material 1-Methyl-3-pyrrolidinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.