missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-piperidinemethanol, 95%, Thermo Scientific™

Catalog Number 11976301
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7583-53-1
C7H15NO
129.2
UGXQXVDTGJCQHR-UHFFFAOYSA-N
1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol,1-methyl-,--9ci
97998
(1-methylpiperidin-3-yl)methanol
CN1CCCC(C1)CO
This item is not returnable. View return policy
7583-53-1
C7H15NO
129.2
UGXQXVDTGJCQHR-UHFFFAOYSA-N
1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol,1-methyl-,--9ci
97998
(1-methylpiperidin-3-yl)methanol
CN1CCCC(C1)CO
CAS Min % 94.0
CAS Max % 100.0
Density 1.0100g/mL
Boiling Point 140.0°C to 145.0°C
Flash Point 94°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4772 to 1.4792
Quantity 50 g
Beilstein 21,II,8
Specific Gravity 1.01
Formula Weight 129.2
Percent Purity 95%
Chemical Name or Material 1-Methyl-3-piperidinemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-3-piperidinemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'