missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenylhydantoin, 95%, Thermo Scientific™

Product Code. 15419798
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2221-12-7
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD00464654
InChI Key YLKKRCYFGKGCKZ-UHFFFAOYSA-N
Synonym 1-methyl-3-phenylhydantoin, 2,4-imidazolidinedione, 1-methyl-3-phenyl
PubChem CID 10774030
IUPAC Name 1-methyl-3-phenylimidazolidine-2,4-dione
SMILES CN1CC(=O)N(C1=O)C2=CC=CC=C2

Specifications

Quantity 5 g
Formula Weight 190.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Methyl-3-phenylhydantoin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'