missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid, 97%, Maybridge™

Brand:  Maybridge CC46601DA

Code : 31

Additional Details : CAS Number : 10250-64-3 Weight : 0.05100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10051054

  • £108.00 / 1g

Estimated Shipment
02-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

10250-64-3
202.213
RTBQZMPYPFQBHA-UHFFFAOYSA-N
7537625
CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O
C11H10N2O2
MFCD08271939
1-methyl-3-phenyl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-phenyl-2h-pyrazole-3-carboxylic acid, 1-methyl-3-phenylpyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl-3-phenyl, 5-carboxy-1-methyl-3-phenyl-1h-pyrazole, 1-methyl-3-phenyl-1h-5-pyrazolecarboxylic acid, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-phenyl, 1-methyl-3-phenyl-1 h-pyrazole-5-carboxylic acid
2-methyl-5-phenylpyrazole-3-carboxylic acid

Specifications

1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
1g
96.5
97%
MFCD08271939
RTBQZMPYPFQBHA-UHFFFAOYSA-N
2-methyl-5-phenylpyrazole-3-carboxylic acid
7537625
97%
Amber Glass Bottle
10250-64-3
100.0
C11H10N2O2
1-methyl-3-phenyl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-phenyl-2h-pyrazole-3-carboxylic acid, 1-methyl-3-phenylpyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl-3-phenyl, 5-carboxy-1-methyl-3-phenyl-1h-pyrazole, 1-methyl-3-phenyl-1h-5-pyrazolecarboxylic acid, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-phenyl, 1-methyl-3-phenyl-1 h-pyrazole-5-carboxylic acid
CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O
202.213
202.21
Videos
SDS Documents