missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. p-2695871
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10242882

Brand: Thermo Scientific Maybridge KM01136DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10199-50-5
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD00067874
InChI Key KCYRMURRLLYLPU-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
PubChem CID 517779
IUPAC Name 2-methyl-5-phenylpyrazol-3-amine
SMILES CN1C(=CC(=N1)C2=CC=CC=C2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazol-5-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.