missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium chloride, 97%, Thermo Scientific Chemicals

Product Code. 15445888
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64697-40-1
Molecular Formula C12H23ClN2
Molecular Weight (g/mol) 230.78
MDL Number MFCD03095432
InChI Key OXFBEEDAZHXDHB-UHFFFAOYSA-M
Synonym 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
PubChem CID 2734223
ChEBI CHEBI:61342
IUPAC Name 1-methyl-3-octylimidazol-1-ium;chloride
SMILES [Cl-].CCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Density 1.01 g/mL
Refractive Index 1.51
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 230.78
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'