missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%

Product Code. 11303558
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303558 4 L 4 L
11383548 1 L 1 L
11393548 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11303558 Supplier Thermo Scientific Alfa Aesar Supplier No. 039176.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For Spectrophotometry/ Chromatography | CAS: 872-50-4 | C5H9NO | 99.13 g/mol

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Evaporation residue 6ppm, H2O 0.05%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point °C to 24°C
Color Colorless
Appearance Colorless liquid
Density 1.033 g/cm3 at 20°C
Quantity 4 L
Flash Point 86°C (187°F)
Ignition Point 270°C (518°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.4705
Vapor Pressure 0.29 hPa
Beilstein 106420
Boiling Point 204°C
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Grade Spectrophotometry
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.