missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Reagent Grade, 99%, Honeywell™

Product Code. 16674464 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16674464 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 16674464 Supplier Honeywell Research Chemicals Supplier No. M796034L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 872-50-4
CAS Max % 1
Melting Point -24°C
Color Colorless
Density 1.028 g/cm3 (20°C)
pH 8.5 to 10
Boiling Point 204°C (1013 hPa)
Flash Point 91°C
Molecular Formula C5H9NO
Linear Formula C5H9NO
Quantity 4 L
Beilstein 106420
Synonym 1-Methyl-2-pyrrolidone; N-Methyl-2-pyrrolidone; NMP
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.13
Grade Reagent Grade Solvents
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.