missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Solstice™

Product Code. 17864591
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
17864591 6 x 1 L Glass Bottle 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17864591 Supplier Chromasolv Supplier No. 693376X1L
Solstice Chromasolv Promotion
Use promo code 27944
View all promotions
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

GC-Headspace tested, ≥99.9%

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Specifications

CAS 872-50-4
Quantity 6 x 1 L
Flash Point 88 °C
Packaging Glass Bottle
Physical Form Liquid, Clear
Molecular Formula C5H9NO
Boiling Point 202°C
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.13
PubChem CID 13387
ChEBI CHEBI:7307
Percent Purity min. 99.9 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.