missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 11360358
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 100 mL
4 x 1 L
Unit Size:
1 L
100mL
4 L
400mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11360358 1 L 1 L
11380358 100 mL 100mL
11350358 4 x 100 mL 400mL
11370358 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11360358 Supplier Thermo Scientific Alfa Aesar Supplier No. 043741.K2
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 872-50-4 | C5H9NO | 99.13 g/mol

1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24°C
Color Colorless to Yellow
Density 1.033 g/mL
Quantity 1 L
Flash Point 86°C (187°F)
Odor Amine-like
Packaging ChemSeal™ Bottle
Physical Form Liquid
Refractive Index 1.4705
Beilstein 106420
Boiling Point 204°C
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Chemical Name or Material 1-Methyl-2-Pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone, anhydrous
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Methyl-2-pyrrolidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.