missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-Pyrrolidinone, 99%, Reagent Grade, Honeywell™

Product Code. p-7120032 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
18 L
25 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
18 L
25 L
Pack of 4
Pack of 6
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15607970

Brand: Honeywell Research Chemicals M796031L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 8.5 to 10 (at 100.00g/L 20°C)
Melting Point -24°C
Color Colorless
Density 1.028g/mL at 25°C
Boiling Point 204°C (1013 hPa)
Quantity 1 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 0.99mmHg (40°C), 0.29mmHg (20°C)
Beilstein 106420
Solubility Information Fully miscible in water
Formula Weight 99.13g/mol
Percent Purity 0.99
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N-Methyl-2-pyrrolidone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.