missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Product Code. p-3881722
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10586894 250 mg Amber Glass Bottle 250mg
10727105 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10586894

Brand: Thermo Scientific Maybridge CC77402CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79583-19-0
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD03421496
InChI Key QLBBQLJPRXPVOS-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl, 1h-pyrazole-4-carbonylchloride, 1-methyl, 1-methyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-9ci
PubChem CID 3164931
IUPAC Name 1-methylpyrazole-4-carbonyl chloride
SMILES CN1C=C(C=N1)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.