missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-boronic acid hydrochloride, 95%, Thermo Scientific™

Product Code. p-4812623 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11986241

Brand: Thermo Scientific Acros 445490010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1026796-02-0
Molecular Formula C4H8BClN2O2
Molecular Weight (g/mol) 162.38
MDL Number MFCD09972098
InChI Key YFPLZINTZVVVJM-UHFFFAOYSA-N
Synonym 1-methylpyrazole-4-boronic acid, hcl, 1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid,hcl, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl
PubChem CID 45158882
IUPAC Name (1-methylpyrazol-4-yl)boronic acid;hydrochloride
SMILES Cl.CN1C=C(C=N1)B(O)O

Specifications

Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.38
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
compliance-icons
Product Identifier
  • 1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.