missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10235313
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235313 1 g Amber Glass Bottle 1g
10757623 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235313 Supplier Thermo Scientific Maybridge Supplier No. CC48303DA
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 89501-90-6
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.606
MDL Number MFCD08690273
InChI Key TWLAHGNFQBQYEL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-sulfonyl chloride, 1-methyl-1h-pyrazole-3-sulfonylchloride, 1h-pyrazole-3-sulfonyl chloride, 1-methyl, 1h-pyrazole-3-sulfonylchloride, 1-methyl, pubchem14132, acmc-209zg6, n-methylpyrazole sulfonyl chloride, chloro 1-methylpyrazol-3-yl sulfone, 1-methyl-3-pyrazolesulfonyl chloride, methyl-1-h-pyrazole-3-sulfonyl chloride
PubChem CID 13474466
IUPAC Name 1-methylpyrazole-3-sulfonyl chloride
SMILES CN1C=CC(=N1)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.