missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-5-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3541109
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
84547-61-5
C5H8N2O
112.132
MFCD08556301
WQFOGLYQVFBDEY-UHFFFAOYSA-N
1-methyl-1h-pyrazol-5-yl methanol, 5-hydroxymethyl-1-methyl-1h-pyrazole, 1h-pyrazole-5-methanol, 1-methyl, 2-methylpyrazol-3-yl methanol, 5-hydroxymethyl-1-methylpyrazole, 1h-pyrazole-5-methanol,1-methyl, 1-methylpyrazol-5-yl methan-1-ol, 1-methylpyrazole-5-methanol, 1-methyl-1h-pyrazol-5-yl ;methanol
11815316
(2-methylpyrazol-3-yl)methanol
CN1C(=CC=N1)CO
This item is not returnable. View return policy
84547-61-5
C5H8N2O
112.132
MFCD08556301
WQFOGLYQVFBDEY-UHFFFAOYSA-N
1-methyl-1h-pyrazol-5-yl methanol, 5-hydroxymethyl-1-methyl-1h-pyrazole, 1h-pyrazole-5-methanol, 1-methyl, 2-methylpyrazol-3-yl methanol, 5-hydroxymethyl-1-methylpyrazole, 1h-pyrazole-5-methanol,1-methyl, 1-methylpyrazol-5-yl methan-1-ol, 1-methylpyrazole-5-methanol, 1-methyl-1h-pyrazol-5-yl ;methanol
11815316
(2-methylpyrazol-3-yl)methanol
CN1C(=CC=N1)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'