missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-3-yl)methanol, 95%, Thermo Scientific™

Product Code. 10122514
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122514 1 g Amber Glass Bottle 1g
10651704 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10122514 Supplier Thermo Scientific Maybridge Supplier No. CC48309DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 84547-62-6
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD08690274
InChI Key WCKJRVKJTUIAFW-UHFFFAOYSA-N
PubChem CID 12845046
IUPAC Name (1-methylpyrazol-3-yl)methanol
SMILES CN1C=CC(=N1)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-methyl-1h-pyrazol-3-yl methanol, 1h-pyrazole-3-methanol, 1-methyl, 1-methylpyrazol-3-yl methanol, 1h-pyrazole-3-methanol,1-methyl, 3-hydroxymethyl-1-methyl-1h-pyrazole, 1-methylpyrazol-3-yl methan-1-ol, akos pao-1076, pubchem23752, 1-methyl-3-pyrazolyl methanol, hydroxymethyl-1-methylpyrazole
Formula Weight 112.13
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-pyrazol-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.