missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazol-3-amine, 97%, Thermo Scientific™

Product Code. 10081784
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10081784 10 g 10g
10344762 250 mg 250mg
10121262 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081784 Supplier Thermo Scientific Maybridge Supplier No. GK00321EA
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1904-31-0
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
InChI Key MOGQNVSKBCVIPW-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole
PubChem CID 137254
IUPAC Name 1-methylpyrazol-3-amine
SMILES CN1C=CC(=N1)N

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazol-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.