missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10739415
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 202745-73-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD08690250
InChI Key DJQOGNYSBOIJKE-UHFFFAOYSA-N
PubChem CID 19819261
IUPAC Name 1-methylindole-6-carboxylic acid
SMILES CN1C=CC2=C1C=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-indole-6-carboxylic acid, 1h-indole-6-carboxylic acid, 1-methyl, 1-methyl-6-indolecarboxylic acid, 6-carboxy-1-methyl-1h-indole, 1-methyl-ih-indole-6-carboxylic acid, 1h-indole-6-carboxylicacid,1-methyl, 1h-indole-6-carboxylicacid, 1-methyl
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'