missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10633635
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10633635

Brand: Thermo Scientific Maybridge CC41401CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 186129-25-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD03839859
InChI Key UHQAIJFIXCOBCN-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-5-indolecarboxylic acid, 1h-indole-5-carboxylic acid, 1-methyl, 1-methyl-1h-indole-5-carboxylicacid, 5-carboxy-1-methyl-1h-indole, 1h-indole-5-carboxylicacid,1-methyl, n-methylindol-5-carboxylic acid, 1-methyl-5-indolecarboxylicacid, methyl 1h-indole-5-carboxylic acid, 1-methyl-1h-indol-5-carboxylic acid
PubChem CID 7015601
IUPAC Name 1-methylindole-5-carboxylic acid
SMILES CN1C=CC2=C1C=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.