missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10321865
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10321865 5 g 5g
10633635 250 mg 250mg
10132804 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10321865 Supplier Thermo Scientific Maybridge Supplier No. CC41401DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 186129-25-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD03839859
InChI Key UHQAIJFIXCOBCN-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-5-indolecarboxylic acid, 1h-indole-5-carboxylic acid, 1-methyl, 1-methyl-1h-indole-5-carboxylicacid, 5-carboxy-1-methyl-1h-indole, 1h-indole-5-carboxylicacid,1-methyl, n-methylindol-5-carboxylic acid, 1-methyl-5-indolecarboxylicacid, methyl 1h-indole-5-carboxylic acid, 1-methyl-1h-indol-5-carboxylic acid
PubChem CID 7015601
IUPAC Name 1-methylindole-5-carboxylic acid
SMILES CN1C=CC2=C1C=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.