missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. p-3175477
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10442284

Brand: Thermo Scientific Maybridge CC41404CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 90923-75-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08271908
InChI Key XIVDZBIBWGQOTI-UHFFFAOYSA-N
PubChem CID 7537534
SMILES CN1C=CC2=CC(C=O)=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 1-methyl, 5-formyl-1-methylindole, n-methylindol-5-carbaldehyde, n-methylindole-5-carbaldehyde, n-methylindole-5-carboxaldehyde, 1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde,1-methyl
IUPAC Name 1-methyl-1H-indole-5-carbaldehyde
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.