missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1H-indole-4-carbonitrile, Thermo Scientific™

Product Code. 10153643
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10153643 1 g 1g
10507494 250 mg 250mg
10290455 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153643 Supplier Thermo Scientific Maybridge Supplier No. CC45816DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 628711-58-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD08690265
InChI Key DYOVHBKIEBCIIE-UHFFFAOYSA-N
PubChem CID 22391592
SMILES CN1C=CC2=C(C=CC=C12)C#N

Specifications

Assay Percent Range 97%
Quantity 1 g
Synonym 1-methyl-1h-indole-4-carbonitrile, 1h-indole-4-carbonitrile,1-methyl, 1h-indole-4-carbonitrile, 1-methyl
IUPAC Name 1-methylindole-4-carbonitrile
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.