missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-3,5,6-triol, TRC

Product Code. 30132722
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132722 100 mg 100mg
30011386 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132722 Supplier Toronto Research Chemicals Supplier No. TRCM313550100MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-75-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.17
Synonym 3,5,6-Trihydroxy-1-methylindole, Adrenolutin, N-Methyl-3,5,6-trihydroxyindole, N-Methyl-5,6-dihydroxyindoxyl;
IUPAC Name 1-methylindole-3,5,6-triol
SMILES OC1=CN(C)C2=C1C=C(O)C(O)=C2>>O=C3CN(C)C4=C3C=C(O)C(O)=C4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H9NO3/c1-10-4-9(13)5-2-7(11)8(12)3-6(5)10/h2-4,11-13H,1H3
Formula Weight 179.0582
Chemical Name or Material 1-Methyl-1H-indole-3,5,6-triol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.