missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-4-boronic acid, 95%, Thermo Scientific™

Product Code. 15294338 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15294338

Brand: Thermo Scientific Acros 454342500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1001907-60-3
Molecular Formula C8H9BN2O2
Molecular Weight (g/mol) 175.98
MDL Number MFCD09870045
InChI Key MDEHELJMJCXYIU-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-4-yl boronic acid, 1-methylindazole-4-boronic acid, 1-methyl-1h-indazole-4-boronic acid, 1-methylindazol-4-boronic acid, 1-methyl-1h-indazol-4-ylboronic acid, 1-methyl-1h-indazol-4-yl-4-boronic acid, acmc-2097nc, 4-borono-1-methyl-1h-indazole, 1-methyl-4-indazolyl boronic acid, 1-methylindazol-4-yl boronic acid
PubChem CID 44118212
SMILES CN1N=CC2=C(C=CC=C12)B(O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
IUPAC Name (1-methyl-1H-indazol-4-yl)boronic acid
Formula Weight 175.98
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indazole-4-boronic acid
compliance-icons
Product Identifier
  • 1-Methyl-1H-indazole-4-boronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.