Learn More
1-methyl-1h-indazole-3-carbonyl chloride, 97%, Maybridge™
Brand: Maybridge CC06302CB
Code : 31
Additional Details : CAS Number : 106649-02-9 Weight : 0.05025kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
106649-02-9 | |
194.618 | |
1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride | |
1-methylindazole-3-carbonyl chloride |
C9H7ClN2O | |
NPNWVMBPSINFBV-UHFFFAOYSA-N | |
7022055 | |
CN1C2=CC=CC=C2C(=N1)C(=O)Cl |
Specifications
1-Methyl-1H-indazole-3-carbonyl chloride | |
250mg | |
97% | |
1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride | |
CN1C2=CC=CC=C2C(=N1)C(=O)Cl | |
194.618 | |
194.62 |
Amber Glass Bottle | |
106649-02-9 | |
C9H7ClN2O | |
NPNWVMBPSINFBV-UHFFFAOYSA-N | |
1-methylindazole-3-carbonyl chloride | |
7022055 | |
97% |