missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carbonyl chloride hydrochloride, Thermo Scientific™

Product Code. 10272782
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10272782 1 g Amber Glass Bottle 1g
10733623 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10272782 Supplier Thermo Scientific Maybridge Supplier No. CC23602DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 343569-06-2
Molecular Formula C5H6Cl2N2O
Molecular Weight (g/mol) 181.02
MDL Number MFCD03659705
InChI Key IRADJQMSOFSMNS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carbonyl chloride hydrochloride, 3-methylimidazole-4-carbonyl chloride hydrochloride, 1-methylimidazole-5-carbonyl chloride hcl, 3-methyl-3h-imidazole-4-carbonyl chloride hydrochloride, 3-methyl-4-imidazolecarbonyl chloride hydrochloride, 5-chlorocarbonyl-1-methyl-1h-imidazole hydrochloride, 1h-imidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
PubChem CID 2776553
SMILES Cl.CN1C=NC=C1C(Cl)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 3-methylimidazole-4-carbonyl chloride;hydrochloride
Formula Weight 181.02
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.