missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-sulfonyl chloride, 95+%, Thermo Scientific™

Product Code. 10618172
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10618172 250 mg Amber Glass Bottle 250mg
10272682 1 g Amber Glass Bottle 1g
10193484 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10618172 Supplier Thermo Scientific Maybridge Supplier No. CC03603CB
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 137049-00-4
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.61
MDL Number MFCD00068060
InChI Key KXUGUWTUFUWYRS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchloride
PubChem CID 2736887
IUPAC Name 1-methylimidazole-4-sulfonyl chloride
SMILES CN1C=NC(=C1)S(Cl)(=O)=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.61
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-imidazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.