missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-sulfonamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl |
Formula Weight | 161.18 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1H-imidazole-4-sulfonamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'