missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-4-yl)methylamine, 95%, Thermo Scientific™

Product Code. 10021034
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021034 5 g Amber Glass Bottle 5g
10423312 250 mg Amber Glass Bottle 250mg
10738534 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021034 Supplier Thermo Scientific Maybridge Supplier No. CC03613DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 486414-83-9
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD06411536
InChI Key YSEAGFBRAQOCFM-UHFFFAOYSA-N
PubChem CID 2795114
IUPAC Name (1-methylimidazol-4-yl)methanamine
SMILES CN1C=NC(CN)=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 1-methyl-1h-imidazol-4-yl methanamine, 1-methyl-1h-imidazol-4-yl methylamine, 1h-imidazole-4-methanamine, 1-methyl, 1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methylimidazole, c-1-methyl-1h-imidazol-4-yl-methylamine, 1-1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methyl-1h-imidazole, 1h-imidazole-4-methanamine,1-methyl, 1-methyl-1h-imidazol-4-yl methyl amine
Formula Weight 111.15
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.