missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10255514
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10255514 5 g 5g
10661293 250 mg 250mg
10009321 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255514 Supplier Thermo Scientific Maybridge Supplier No. CC23504DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-67-1
Molecular Formula C8H7N3O
Molecular Weight (g/mol) 161.164
InChI Key IPVPVOVAJDRRAK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzo d 1,2,3 triazole-5-carbaldehyde, 1-methyl-1h-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-1h-benzotriazole-5-carbaldehyde, 1h-benzotriazole-5-carboxaldehyde, 1-methyl, 1-methyl-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-5-benzotriazolecarboxaldehyde, 1-methylbenzotriazole-5-carboxaldehyde
PubChem CID 2795417
IUPAC Name 1-methylbenzotriazole-5-carbaldehyde
SMILES CN1C2=C(C=C(C=C2)C=O)N=N1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.