missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-phenylhydrazine, 97%

Product Code. 11420587
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420587 5 g 5g
11430587 25 g 25g
11440587 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420587 Supplier Thermo Scientific Alfa Aesar Supplier No. B22440.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-40-6 | C7H10N2 | 122.171 g/mol

1-Methyl-1-phenylhydrazine is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-1-phenylhydrazine is used to make other chemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-40-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00007627
InChI Key MWOODERJGVWYJE-UHFFFAOYSA-N
Synonym 1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
PubChem CID 36299
IUPAC Name 1-methyl-1-phenylhydrazine
SMILES CN(C1=CC=CC=C1)N

Specifications

Density 1.038
Boiling Point 54°C to 55°C (0.3 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.583
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 1-Methyl-1-phenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.