Learn More
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Especificaciones
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Color | Colorless |
| Density | 0.7800g/mL |
| Boiling Point | 72°C (754.0 mmHg) |
| Flash Point | -17°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% |
| Packaging | Glass bottle |
| Linear Formula | C5H7CH3 |
| Mostrar más |
Seguridad y manipulación
- 1-Methyl-1-cyclopentene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H304-May be fatal if swallowed and enters airways.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.