missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 15441289
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4965-09-7
MFCD00798989
1-methyl-1,2,3,4-tetrahydroisoquinoline
This item is not returnable. View return policy
4965-09-7
MFCD00798989
1-methyl-1,2,3,4-tetrahydroisoquinoline

1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity. Following systemic administration in rats, 1-Methyl-1,2,3,4-tetrahydroisoquinoline produces antidepressant-like effect similar to the effect of imipramine. 1MeTIQ is a reversible short-acting moderate inhibitor of MAO A/B. Also known as 1,2,3,4-Tetrahydro-1-methylisoquinoline, 1MeTIQ. It is an important raw material and intermediate for revaprazan and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Boiling Point 119°C (18 mmHg)
Molecular Formula C10H13N
Quantity 1 g
Beilstein 82140
Synonym 1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
Solubility Information Slightly soluble in water.
InChI Key QPILYVQSKNWRDD-UHFFFAOYNA-N
SMILES CC1C2=CC=CC=C2CCN1
Molecular Weight (g/mol) 147.22
PubChem CID 92214
Formula Weight 147.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1,2,3,4-tetrahydroisoquinoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'