missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxynaphthalene, TRC

Product Code. 30008683
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008683 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008683 Supplier Toronto Research Chemicals Supplier No. TRCM2272501G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2216-69-5
Molecular Formula C11 H10 O
Quantity 1 g
Synonym 1-Naphthol Methyl Ether, 1-Naphthyl Methyl Ether, Methyl 1-Naphthyl Ether, NSC 5530, α-Methoxynaphthalene, α-Naphthyl Methyl Ether
InChI Formula InChI=1S/C11H10O/c1-12-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3
SMILES COc1cccc2ccccc12
IUPAC Name 1-methoxynaphthalene
Molecular Weight (g/mol) 158.2
Formula Weight 158.0732
Chemical Name or Material 1-Methoxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.