missing translation for 'onlineSavingsMsg'
Learn More

1-(Methoxycarbonyl-ethyl)-4-[(2-methyl-3-oxo-pentanoyl)-phenyl-amino]-piperidine -4-carboxylic Acid Methyl Ester, TRC

Product Code. 30483506
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483506 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483506 Supplier Toronto Research Chemicals Supplier No. TRCM3293865MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Methoxycarbonyl-ethyl)-4-[(2-methyl-3-oxo-pentanoyl)-phenyl-amino]-piperidine-4-carboxylic Acid Methyl Ester
CAS 2518277-19-3
Molecular Formula C23H32N2O6
Quantity 5 mg
Synonym Remifentanil Impurity E, Methyl 1-(3-methoxy-3-oxopropyl)-4-(2-methyl-3-oxo-N- phenylpentanamido)piperidine-4-carboxylate
InChI Formula InChI=1S/C23H32N2O6/c1-5-19(26)17(2)21(28)25(18-9-7-6-8-10-18)23(22(29)31-4)12-15-24(16-13-23)14-11-20(27)30-3/h6-10,17H,5,11-16H2,1-4H3
SMILES CC(C(CC)=O)C(N(C1(C(OC)=O)CCN(CCC(OC)=O)CC1)C2=CC=CC=C2)=O
IUPAC Name methyl 1-(3-methoxy-3-oxopropyl)-4-(2-methyl-3-oxo-N-phenylpentanamido)piperidine-4-carboxylate; 1-Piperidinepropanoic acid, 4-(methoxycarbonyl)-4-[(2-methyl-1,3-dioxopentyl)phenylamino]-, methyl ester;
Molecular Weight (g/mol) 432.51
Formula Weight 432.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.