missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-Empagliflozin Tetraacetoxy, TRC

Product Code. 30386224
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386224 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386224 Supplier Toronto Research Chemicals Supplier No. TRCE52116010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Empagliflozin Tetraacetoxy
CAS 1620758-21-5
Molecular Formula C32 H37 Cl O12
Quantity 10 mg
Synonym Empagliflozin Tetraacetoxy, α-D-Glucopyranoside, methyl 1-C-[4-chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-, 2,3,4,6-tetraacetate (ACI), Methyl 1-C-[4-chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-α-D-glucopyranoside 2,3,4,6-tetraacetate
InChI Formula InChI=1S/C32H37ClO12/c1-18(34)40-17-28-29(41-19(2)35)30(42-20(3)36)31(43-21(4)37)32(38-5,45-28)24-8-11-27(33)23(15-24)14-22-6-9-25(10-7-22)44-26-12-13-39-16-26/h6-11,15,26,28-31H,12-14,16-17H2,1-5H3/t26-,28+,29+,30-,31+,32-/m0/s1
SMILES CO[C@]1(O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)c2ccc(Cl)c(Cc3ccc(O[C@H]4CCOC4)cc3)c2
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[4-chloro-3-[[4-[(3S)-oxolan-3-yl]oxyphenyl]methyl]phenyl]-6-methoxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 649.082
Formula Weight 648.197
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.