missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy Amiodarone, TRC

Product Code. 30475311
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475311 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475311 Supplier Toronto Research Chemicals Supplier No. TRCM26091510MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methoxy Amiodarone
CAS 1087223-70-8
Molecular Formula C26 H31 I2 N O4
Quantity 10 mg
Synonym [4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl][2-[(1RS)-1-methoxybutyl]benzofuran-3-yl]methanone, Amiodarone Hydrochoride Imp. G (EP), 1-Methoxy Amiodarone, [2-[(1RS)-1-Methoxybutyl]benzofuran-3-yl][4-[2-(diethyl-amino)ethoxy]-3,5-diiodophenyl]methanone
InChI Formula InChI=1S/C26H31I2NO4/c1-5-10-22(31-4)26-23(18-11-8-9-12-21(18)33-26)24(30)17-15-19(27)25(20(28)16-17)32-14-13-29(6-2)7-3/h8-9,11-12,15-16,22H,5-7,10,13-14H2,1-4H3
SMILES CCCC(OC)c1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3
IUPAC Name [4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]-[2-(1-methoxybutyl)-1-benzofuran-3-yl]methanone
Molecular Weight (g/mol) 675.34
Formula Weight 675.0342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.