missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene, 90%

Product Code. 10206930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206930 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206930 Supplier Thermo Scientific Acros Supplier No. 199550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59414-23-2 | C8H16O2Si | 172.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59414-23-2
CAS Min % 88.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.8800g/mL
Boiling Point 68.0°C to 69.0°C (14.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C8H16O2Si
Linear Formula (CH3)3SiOC(=CH2)CH=CHOCH3
Refractive Index 1.4530 to 1.4550
MDL Number MFCD00008498
Quantity 5 g
Fieser 06,370; 07,233; 09,303; 10,260; 11,332
Merck Index 15, 6075
Specific Gravity 0.88
Synonym unii-w961bik2c3, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene, silane, 3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, methyl 3-trimethylsilyloxybuta-1,3-dien-1-yl ether, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, z, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene unspecified, silane, 2z-3-methoxy-1-methylene-2-propen-1-yl oxy trimethyl, silane, 3-methoxy-1-methylene-2-propenyl oxy trimethyl-, z
InChI Key SHALBPKEGDBVKK-SREVYHEPSA-N
SMILES COC=CC(=C)O[Si](C)(C)C
IUPAC Name [(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
Molecular Weight (g/mol) 172.3
PubChem CID 6436851
Formula Weight 172.3
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-3-(trimethylsilyloxy)-1, 3-butadiene, 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.