missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-3-trimethylsiloxy-1,3-butadiene, 96%

Product Code. 11333646
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333646 5 g 5g
11323646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333646 Supplier Thermo Scientific Alfa Aesar Supplier No. L14672.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54125-02-9 | C8H16O2Si | 172.299 g/mol

1-Methoxy-3-trimethylsiloxy-1,3-butadiene is employed as a reagent in Mannich-Michael reaction for the synthesis of piperidinones and enaminones. As Diels-Alder diene, it is used in the synthesis of pyridones and pyranones, sulfone analogues of griseofulvin (sulfogriseofulvins) and 4H-1-aminopyrroles and 4, 5H-pyrazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methoxy-3-trimethylsiloxy-1,3-butadiene is employed as a reagent in Mannich-Michael reaction for the synthesis of piperidinones and enaminones. As Diels-Alder diene, it is used in the synthesis of pyridones and pyranones, sulfone analogues of griseofulvin (sulfogriseofulvins) and 4H-1-aminopyrroles and 4, 5H-pyrazoles.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Moisture sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54125-02-9
Molecular Formula C8H16O2Si
Molecular Weight (g/mol) 172.299
MDL Number MFCD00008498
InChI Key SHALBPKEGDBVKK-VOTSOKGWSA-N
Synonym danishefsky's diene, 1-methoxy-3-trimethylsiloxy-1,3-butadiene, unii-1zor63u51o, 1-methoxy-3-trimethylsiloxy butadiene, 1-methoxy-3-trimethylsilyloxy-1,3-butadiene, e-4-methoxybuta-1,3-dien-2-yl oxy trimethylsilane, e-4-methoxybuta-1,3-dien-2-yloxy trimethylsilane, 1-methoxy-3-trimethylsilyloxy butadiene, silane, 2e-3-methoxy-1-methylene-2-propenyl oxy trimethyl, e-1-methoxy-3-trimethylsilyloxy-1,3-butadiene
PubChem CID 5366448
IUPAC Name [(3E)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
SMILES COC=CC(=C)O[Si](C)(C)C

Specifications

Boiling Point 68°C to 69°C (14mmHg)
Quantity 5 g
UN Number UN1993
Beilstein 1616761
Flash Point 45°C (113°F)
Merck Index 14,6001
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 172.3
Percent Purity 97%
Refractive Index 1.455
Sensitivity Air and moisture sensitive
Density 0.89
Chemical Name or Material 1-Methoxy-3-trimethylsiloxy-1,3-butadiene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.