missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-methyl-2-propanol, 98%, AcroSeal™, Thermo Scientific™

Product Code. 10007859 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10007859 100 mL AcroSeal glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10007859 Supplier Thermo Scientific Acros Supplier No. 381711000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3587-64-2
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8970g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal glass Bottle
Molecular Formula C5H12O2
Linear Formula CH3OCH2C(CH3)2OH
Refractive Index 1.4025 to 1.4065 (20°C, 589nm)
MDL Number MFCD03701584
Quantity 100 mL
Specific Gravity 0.897
Synonym 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol #
Solubility Information Solubility in water: miscible
InChI Key MXUXZWFVAPTPAG-UHFFFAOYSA-N
SMILES COCC(C)(C)O
IUPAC Name 1-methoxy-2-methylpropan-2-ol
Molecular Weight (g/mol) 104.15
PubChem CID 77137
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-methyl-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methoxy-2-methyl-2-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.