missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, 97%

Product Code. 11337056
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337056 5 g 5g
11347056 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337056 Supplier Thermo Scientific Alfa Aesar Supplier No. L06931.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31469-15-5 | C8H18O2Si | 174.32 g/mol

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agents and Strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31469-15-5
Molecular Formula C8H18O2Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00010232
InChI Key JNOGVQJEBGEKMG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-1-trimethylsiloxy propene, methyl trimethylsilyl dimethylketene acetal, dimethylketene methyl trimethylsilyl acetal, 1-methoxy-2-methylprop-1-en-1-yl oxy trimethylsilane, silane, 1-methoxy-2-methyl-1-propenyl oxy trimethyl, 1-methoxy-1-trimethylsiloxy-2-methyl-1-propene, dimethylketene methyltrimethylsilylacetal, 1-methoxy-1-trimethylsilyloxy-2-methylpropene, 1-methoxy-2-methyl-1-propenyloxy trimethylsilane, 1-methoxy-2-methylprop-1-enoxy-trimethylsilane
PubChem CID 552336
IUPAC Name (1-methoxy-2-methylprop-1-enoxy)-trimethylsilane
SMILES COC(O[Si](C)(C)C)=C(C)C

Specifications

Boiling Point 35°C to 36°C (15mmHg)
Quantity 5 g
UN Number UN1993
Beilstein 1362893
Flash Point 30°C (86°F)
Solubility Information Hydrolyzes in water.
Formula Weight 174.32
Percent Purity 97%
Refractive Index 1.415
Sensitivity Moisture sensitive
Density 0.858
Chemical Name or Material 1-Methoxy-2-methyl-1-(trimethylsiloxy)propene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.