Learn More
Fisher Chemical 1 KG SODIUM TETRABORATE (BORAX)
Brand: Fisher Chemical BP/S370/60
1 KG SODIUM TETRABORATE (BORAX)
Description
Safety and Handling
- di-Sodium tetraborate BP
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.