missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropylamino-3-(p-tolyloxy)-2-propanol, TRC

Product Code. 30389262
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389262 50 mg -
30394182 10 mg -
30407795 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389262 Supplier Toronto Research Chemicals Supplier No. TRCI82399550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5790-46-5
Molecular Formula C13 H21 N O2
Molecular Weight (g/mol) 223.31
Synonym (2RS)-1-(Isopropylamino)-3-(4-methylphenoxy)propan-2-ol, Bisoprolol Fumarate Imp. R (EP), 2-Propanol, 1-[(1-methylethyl)amino]-3-(4-methylphenoxy)-, 2-Propanol, 1-(isopropylamino)-3-(p-tolyloxy)- (7CI,8CI), 1-[(1-Methylethyl)amino]-3-(4-methylphenoxy)-2-propanol, 1-(4-Methylphenoxy)-3-[(propan-2-yl)amino]propan-2-ol, 1-Isopropylamino-3-(p-tolyloxy)-2-propanol, [2-Hydroxy-3-(4-methylphenoxy)propyl](propan-2-yl)amine
IUPAC Name 1-(4-methylphenoxy)-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(O)COc1ccc(C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H21NO2/c1-10(2)14-8-12(15)9-16-13-6-4-11(3)5-7-13/h4-7,10,12,14-15H,8-9H2,1-3H3
Formula Weight 223.1572
Chemical Name or Material (2RS)-1-(Isopropylamino)-3-(4-methylphenoxy)propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.