missing translation for 'onlineSavingsMsg'
Learn More

1-Isocyanato-3-phenoxybenzene, TRC

Product Code. 30400185
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400185 1 g
30408697 100 mg
30391064 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400185 Supplier Toronto Research Chemicals Supplier No. TRCI8091601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87873-72-1
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.21
Synonym 3-Phenoxyphenyl Isocyanate, m-Phenoxyphenylisocyanate
SMILES O=C=Nc1cccc(Oc2ccccc2)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H9NO2/c15-10-14-11-5-4-8-13(9-11)16-12-6-2-1-3-7-12/h1-9H
Formula Weight 211.0633
Chemical Name or Material 1-Isocyanato-3-phenoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.