missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane, TRC

Product Code. 30398066
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398066 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398066 Supplier Toronto Research Chemicals Supplier No. TRCI7174535ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Iodopentane
CAS 628-17-1
Molecular Formula C5H11I
Quantity 5 mL
Synonym Pentane, 1-iodo- (6CI, 8CI, 9CI, ACI), 1-Iodopentane (ACI), 1-Pentyl iodide, 5-Iodopentane, Amyl iodide, n-Amyl iodide, n-Pentyl iodide, NSC 7901, Pentyl iodide
InChI Formula InChI=1S/C5H11I/c1-2-3-4-5-6/h2-5H2,1H3
SMILES CCCCCI
IUPAC Name 1-iodopentane
Molecular Weight (g/mol) 198.04
Formula Weight 197.9905
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.